Gene Transcripts

Gene Symbol
OR52I1
Entrez Gene ID
390037
Gene Name
olfactory receptor family 52 subfamily I member 1
Gene Aliases
OR11-13
Chromosome Location
Chr. 11: 4594039 - 4595013 on Build GRCh38
UniGene ID
Hs.553727
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005169.1 NP_001005169.1 1 466

View Full Product Details