Gene Transcripts

Gene Symbol
OR9G1
Entrez Gene ID
390174
Gene Name
olfactory receptor family 9 subfamily G member 1
Gene Aliases
OR9G5
Chromosome Location
Chr. 11: 56700388 - 56701305 on Build GRCh38
UniGene ID
Hs.553752
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005213.1 NP_001005213.1 1 157

View Full Product Details