Gene Transcripts

Gene Symbol
ANKRD20A3
Entrez Gene ID
441425
Gene Name
ankyrin repeat domain 20 family member A3
Gene Aliases
Chromosome Location
Chr. 9: 66093946 - 66153193 on Build GRCh38
UniGene ID
Hs.632663
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012419.2 NP_001012419.1 15 3492

Gene Symbol
ANKRD20A8P
Entrez Gene ID
729171
Gene Name
ankyrin repeat domain 20 family member A8, pseudogene
Gene Aliases
ANKRD20B
Chromosome Location
Chr. 2: 94760928 - 94857075 on Build GRCh38
UniGene ID
Hs.456852
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA CR936649.1 - 2 2546

View Full Product Details