Gene Transcripts

Gene Symbol
SPRR2G
Entrez Gene ID
6706
Gene Name
small proline rich protein 2G
Gene Aliases
Chromosome Location
Chr. 1: 153149582 - 153195377 on Build GRCh38
UniGene ID
Hs.490253
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014291.3 NP_001014313.1 2 536
XM_017002177.1 XP_016857666.1 4 913
XM_017002178.1 XP_016857667.1 2 459

View Full Product Details