Gene Transcripts
- Gene Symbol
- MAJIN
- Entrez Gene ID
- 283129
- Gene Name
- membrane anchored junction protein
- Gene Aliases
- C11orf85
- Chromosome Location
- Chr. 11: 64938230 - 64972104 on Build GRCh38
- UniGene ID
- Hs.567793
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001037225.2 |
NP_001032302.1
|
3, 4 |
390 |
|
NM_001300803.1 |
NP_001287732.1
|
3, 4 |
390 |
|
NM_001318808.1 |
NP_001305737.1
|
3, 4 |
390 |
|
XM_005273918.2 |
XP_005273975.1
|
3, 4 |
384 |
|
XM_006718502.2 |
XP_006718565.1
|
3, 4 |
384 |
|
XM_006718503.3 |
XP_006718566.1
|
3, 4 |
270 |
|
XM_006718504.2 |
XP_006718567.1
|
3, 4 |
384 |
|
XM_006718505.2 |
XP_006718568.1
|
3, 4 |
384 |
|
XM_011544940.2 |
XP_011543242.1
|
3, 4 |
265 |
|
GenBank mRNA |
AK093130.1 |
- |
3, 4 |
384 |
|
BC033501.1 |
- |
3, 4 |
322 |
|
BC106951.2 |
- |
2, 3 |
185 |
40035116 |
BC106952.1 |
- |
2, 3 |
185 |
40035117 |
DB445650.2 |
- |
3, 4 |
391 |
|
LC068587.1 |
- |
1, 2 |
111 |
|
View Full Product Details