Gene Transcripts

Gene Symbol
INSC
Entrez Gene ID
387755
Gene Name
inscuteable homolog (Drosophila)
Gene Aliases
Chromosome Location
Chr. 11: 15111416 - 15269675 on Build GRCh38
UniGene ID
Hs.591997
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031853.4 NP_001027024.3 3, 4 585
NM_001042536.2 NP_001036001.1 3, 4 509
NM_001278313.1 NP_001265242.1 2, 3 471
NM_001278314.1 NP_001265243.1 2, 3 471
NM_001278315.1 NP_001265244.1 2, 3 471
NM_001278316.1 NP_001265245.1 2, 3 471
XM_006718227.3 XP_006718290.1 4, 5 638
XM_011520085.1 XP_011518387.1 3, 4 513
XM_011520086.1 XP_011518388.1 3, 4 513
XM_017017697.1 XP_016873186.1 4, 5 550
XM_017017698.1 XP_016873187.1 3, 4 513
GenBank mRNA AB231745.1 - 2, 3 465
AB231746.1 - 2, 3 465
AB231747.1 - 2, 3 465
AB231749.1 - 1, 2 212
AB236158.1 - 3, 4 509
AB236159.1 - 3, 4 585
BC127700.1 - 2, 3 471 40127962
BU147420.1 - 3, 4 496

View Full Product Details