Gene Transcripts
- Gene Symbol
- NBPF8
- Entrez Gene ID
- 728841
- Gene Name
- neuroblastoma breakpoint family member 8
- Gene Aliases
- NBPF8P
- Chromosome Location
- Chr. 1: 120436351 - 120467844 on Build GRCh38
- UniGene ID
- Hs.445080
- Species
- Homo sapiens
- Gene Symbol
- NBPF1
- Entrez Gene ID
- 55672
- Gene Name
- neuroblastoma breakpoint family member 1
- Gene Aliases
- AB13, AB14, AB23, AD2, NBG, NBPF
- Chromosome Location
- Chr. 1: 16562427 - 16613605 on Build GRCh38
- UniGene ID
- Hs.467587
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC094705.1 |
- |
14 |
1812 |
|
View Full Product Details