Gene Transcripts

Gene Symbol
Ugt2b37
Entrez Gene ID
29623
Gene Name
UDP-glucuronosyltransferase 2 family, member 37
Gene Aliases
Chromosome Location
Chr. 14: 22517737 - 22535098 on Build Rnor_6.0
UniGene ID
Rn.230458
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770015.1 XP_008768237.1 6 1757
GenBank mRNA M33746.1 - 6 1736
M33747.1 - 564

View Full Product Details