Gene Transcripts

Gene Symbol
P3h2
Entrez Gene ID
288016
Gene Name
prolyl 3-hydroxylase 2
Gene Aliases
Leprel1
Chromosome Location
Chr. 11: 78028885 - 78169746 on Build Rnor_6.0
UniGene ID
Rn.213595
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025627.1 NP_001020798.1 5, 6 1244
GenBank mRNA BC099107.1 - 5, 6 1244

View Full Product Details