Gene Transcripts

Gene Symbol
Psg29
Entrez Gene ID
292666
Gene Name
pregnancy-specific glycoprotein 29
Gene Aliases
Chromosome Location
Chr. 1: 79236391 - 79259492 on Build Rnor_6.0
UniGene ID
Rn.154014
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025641.1 NP_001020812.1 3 876
GenBank mRNA BC097354.1 - 3 876

View Full Product Details