Gene Transcripts

Gene Symbol
Ceacam11
Entrez Gene ID
292668
Gene Name
carcinoembryonic antigen-related cell adhesion molecule 11
Gene Aliases
Chromosome Location
Chr. 1: 79396578 - 79402814 on Build Rnor_6.0
UniGene ID
Rn.205326
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025404.1 NP_001020575.1 3, 4 806
GenBank mRNA BC097990.1 - 3, 4 806

View Full Product Details