Gene Transcripts

Gene Symbol
Palmd
Entrez Gene ID
310811
Gene Name
palmdelphin
Gene Aliases
Chromosome Location
Chr. 2: 220481292 - 220536004 on Build Rnor_6.0
UniGene ID
Rn.24602
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025688.1 NP_001020859.1 543
GenBank mRNA BC098755.1 - 543

View Full Product Details