Gene Transcripts

Gene Symbol
Colec12
Entrez Gene ID
361289
Gene Name
collectin sub-family member 12
Gene Aliases
Chromosome Location
Chr. 18: 867048 - 1052608 on Build Rnor_6.0
UniGene ID
Rn.31273
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025721.1 NP_001020892.1 5 589
GenBank mRNA AB080968.1 - 1 88
BC097495.1 - 5 589 7461783

View Full Product Details