Gene Transcripts

Gene Symbol
Rhox2
Entrez Gene ID
363439
Gene Name
reproductive homeobox on X chromosome 2
Gene Aliases
Rhox3
Chromosome Location
Chr. X: 123888033 - 123894411 on Build Rnor_6.0
UniGene ID
Rn.220867
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025746.3 NP_001020917.3 2 1151
GenBank mRNA DQ058650.1 - 2 1151

View Full Product Details