Gene Transcripts

Gene Symbol
Yipf6
Entrez Gene ID
363476
Gene Name
Yip1 domain family, member 6
Gene Aliases
Chromosome Location
Chr. X: 68625771 - 68637780 on Build Rnor_6.0
UniGene ID
Rn.101396
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025747.1 NP_001020918.1 7 761
XM_006257077.2 XP_006257139.1 9 918
GenBank mRNA BC097987.1 - 7 761
FQ211792.1 - 7 846

View Full Product Details