Gene Transcripts

Gene Symbol
Syt1
Entrez Gene ID
25716
Gene Name
synaptotagmin I
Gene Aliases
P65
Chromosome Location
Chr. 7: 50084063 - 50638798 on Build Rnor_6.0
UniGene ID
Rn.216272
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033680.2 NP_001028852.2 7 944
XM_008765352.1 XP_008763574.1 4 926
XM_008765353.1 XP_008763575.1 4 920
GenBank mRNA AJ617615.1 - 7 944
CB731032.1 - 3 317
DQ181550.2 - 4 612
X52772.1 - 7 1127

View Full Product Details