Gene Transcripts

Gene Symbol
Dars2
Entrez Gene ID
304919
Gene Name
aspartyl-tRNA synthetase 2 (mitochondrial)
Gene Aliases
RGD1308286
Chromosome Location
Chr. 13: 78857638 - 78885464 on Build Rnor_6.0
UniGene ID
Rn.137838
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034143.1 NP_001029315.1 4, 5 499
GenBank mRNA BC105774.1 - 4, 5 499

View Full Product Details