Gene Transcripts

Gene Symbol
Dctn5
Entrez Gene ID
308961
Gene Name
dynactin 5 (p25)
Gene Aliases
RGD1305654
Chromosome Location
Chr. 1: 192088709 - 192105421 on Build Rnor_6.0
UniGene ID
Rn.25812
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037778.1 NP_001032867.1 6 1340
GenBank mRNA BC098764.1 - 6 1340

View Full Product Details