Gene Transcripts

Gene Symbol
Itgal
Entrez Gene ID
308995
Gene Name
integrin, alpha L
Gene Aliases
LFA-1
Chromosome Location
Chr. 1: 198744051 - 198781745 on Build Rnor_6.0
UniGene ID
Rn.14655
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033998.2 NP_001029170.1 9 992
XM_006230269.2 XP_006230331.1 9 1133
GenBank mRNA AY256462.1 - 4 495
BC101849.1 - 9 992 7938065

View Full Product Details