Gene Transcripts

Gene Symbol
Crtc2
Entrez Gene ID
310615
Gene Name
CREB regulated transcription coactivator 2
Gene Aliases
RGD1308903, Torc2
Chromosome Location
Chr. 2: 189615940 - 189626075 on Build Rnor_6.0
UniGene ID
Rn.13599
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033895.1 NP_001029067.1 13 1743
XM_008761146.1 XP_008759368.1 12 1788
GenBank mRNA BC128708.1 - 13 1868
BC168242.1 - 13 1856 7934746
DQ185515.1 - 13 1743

View Full Product Details