Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001037654.1 | NP_001032743.1 | 14 | 1401 | |
XM_006243042.2 | XP_006243104.1 | 14 | 1858 | ||
XM_006243045.2 | XP_006243107.1 | 13 | 1213 | ||
XM_008766295.1 | XP_008764517.1 | 14 | 1894 | ||
XM_008766296.1 | XP_008764518.1 | 10 | 1225 | ||
GenBank mRNA | AY770507.1 | - | 14 | 1401 |