Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001030044.1 | NP_001025215.1 | 2 | 463 | |
XM_006255197.2 | XP_006255259.1 | 2 | 246 | ||
XM_008772318.1 | XP_008770540.1 | 2 | 235 | ||
GenBank mRNA | DQ109812.1 | - | 2 | 463 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001107413.1 | NP_001100883.1 | 1 | 241 | |
XM_006255191.2 | XP_006255253.1 | 2 | 1220 | ||
XM_006255192.2 | XP_006255254.1 | 2 | 1220 |