Gene Transcripts

Gene Symbol
Gp9
Entrez Gene ID
502858
Gene Name
glycoprotein IX (platelet)
Gene Aliases
Chromosome Location
Chr. 4: 119680671 - 119682006 on Build Rnor_6.0
UniGene ID
Rn.9617
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031825.1 NP_001026995.1 2 220
GenBank mRNA AB158423.1 - 2 220
AB158424.1 - 2 220

View Full Product Details