Gene Transcripts

Gene Symbol
Defa8
Entrez Gene ID
613224
Gene Name
defensin alpha 8
Gene Aliases
Chromosome Location
Chr. 16: 75428675 - 75429568 on Build Rnor_6.0
UniGene ID
Rn.234253
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033077.1 NP_001028249.1 2 261
GenBank mRNA AY623752.1 - 2 261

View Full Product Details