Gene Transcripts

Gene Symbol
CIAPIN1
Entrez Gene ID
57019
Gene Name
cytokine induced apoptosis inhibitor 1
Gene Aliases
Anamorsin, DRE2, PRO0915
Chromosome Location
Chr. 16: 57428169 - 57447528 on Build GRCh38
UniGene ID
Hs.4900
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308347.1 NP_001295276.1 2 329
NM_001308358.1 NP_001295287.1 2 329
NM_020313.3 NP_064709.2 2 329
XM_005256063.4 XP_005256120.1 2 220
GenBank mRNA AF248964.1 - 2 233
AK074263.1 - 2 170
AK292281.1 - 2 172
AK295024.1 - 1 562
AL136613.1 - 2 190
BC002568.2 - 2 171 3139226
BC024196.2 - 2 210 4816594
BC067303.1 - 2 329 30389410
BC071740.1 - 2 204 5563094
BG746807.1 - 1 175
BJ991015.1 - 2 210
CR533545.1 - 1 87

View Full Product Details