Gene Transcripts

Gene Symbol
Scgb1b27
Entrez Gene ID
11354
Gene Name
secretoglobin, family 1B, member 27
Gene Aliases
Abp, Abpa, Abpa27, Sal-1, Tcp
Chromosome Location
Chr. 7: 34021567 - 34022881 on Build GRCm38
UniGene ID
Mm.8025
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009596.1 NP_033726.1 3 398
GenBank mRNA BC016132.1 - 3 398

View Full Product Details