Gene Transcripts

Gene Symbol
PDSS2
Entrez Gene ID
57107
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 2
Gene Aliases
C6orf210, COQ10D3, DLP1, bA59I9.3, hDLP1
Chromosome Location
Chr. 6: 107152557 - 107459683 on Build GRCh38
UniGene ID
Hs.745008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020381.3 NP_065114.3 3, 4 928
XM_011535956.2 XP_011534258.1 3, 4 1026
XM_011535957.2 XP_011534259.1 3, 4 1036
XM_011535958.2 XP_011534260.1 2, 3 892
XM_011535959.2 XP_011534261.1 3, 4 1001
XM_011535960.2 XP_011534262.1 3, 4 655
XM_011535961.2 XP_011534263.1 3, 4 948
XM_011535962.2 XP_011534264.1 3, 4 570
XM_017011082.1 XP_016866571.1 3, 4 948
GenBank mRNA AB210839.1 - 3, 4 638
AF254956.1 - 3, 4 899
AK225629.1 - 3, 4 909
AK296717.1 - 3, 4 909
AK301480.1 - 3, 4 531
AK315804.1 - 3, 4 909
BC039906.1 - 3, 4 914 6066481

View Full Product Details