Gene Transcripts

Gene Symbol
Olfr98
Entrez Gene ID
258503
Gene Name
olfactory receptor 98
Gene Aliases
MOR156-4
Chromosome Location
Chr. 17: 37262733 - 37263662 on Build GRCm38
UniGene ID
Mm.377540
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146510.1 NP_666721.1 1 708
GenBank mRNA BC119060.1 - 1 795
BC120744.1 - 1 795 40129667

View Full Product Details