Gene Transcripts

Gene Symbol
Olfr24
Entrez Gene ID
18322
Gene Name
olfactory receptor 24
Gene Aliases
MOR132-1, MTPCR51
Chromosome Location
Chr. 9: 18754692 - 18755633 on Build GRCm38
UniGene ID
Mm.246530
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146606.1 NP_666817.1 1 926
GenBank mRNA BC125621.1 - 1 1002
BC137822.1 - 1 1002 8860841

View Full Product Details