Gene Transcripts

Gene Symbol
Cyp2c54
Entrez Gene ID
404195
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 54
Gene Aliases
EG404195
Chromosome Location
Chr. 19: 40037940 - 40073845 on Build GRCm38
UniGene ID
Mm.379575
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206537.2 NP_996260.1 3 378
GenBank mRNA AI181879.1 - 3 367
BC043929.1 - 2 160
BF659738.1 - 3 378

View Full Product Details