Gene Transcripts

Gene Symbol
PRTFDC1
Entrez Gene ID
56952
Gene Name
phosphoribosyl transferase domain containing 1
Gene Aliases
HHGP
Chromosome Location
Chr. 10: 24848607 - 24952644 on Build GRCh38
UniGene ID
Hs.405619
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282786.1 NP_001269715.1 3 306
NM_020200.6 NP_064585.1 3 306
GenBank mRNA AB209792.1 - 3 256
AF226056.1 - 3 254
AK021950.1 - 3 266
AK222674.1 - 3 265
AK294130.1 - 3 265
AK304055.1 - 3 265
AK309614.1 - 3 295
BC008662.2 - 3 268 3871911

View Full Product Details