Gene Transcripts
- Gene Symbol
- Mrgpra6
- Entrez Gene ID
- 381886
- Gene Name
- MAS-related GPR, member A6
- Gene Aliases
- MrgA6
- Chromosome Location
- Chr. 7: 47185766 - 47189355 on Build GRCm38
- UniGene ID
- Mm.377894
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC132198.1 |
- |
1 |
86 |
|
BC132204.1 |
- |
1 |
86 |
40130481 |
View Full Product Details