Gene Transcripts

Gene Symbol
Spink7
Entrez Gene ID
408198
Gene Name
serine peptidase inhibitor, Kazal type 7 (putative)
Gene Aliases
EG408198, Ecg2
Chromosome Location
Chr. 18: 62592413 - 62596264 on Build GRCm38
UniGene ID
Mm.478742
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001803.2 NP_001001803.2 4 226

View Full Product Details