Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001009818.1 | NP_001009818.1 | 10 | 1972 | |
GenBank mRNA | AK136162.1 | - | 4 | 957 | |
AK166517.1 | - | 10 | 1994 | ||
AK170945.1 | - | 9 | 1782 | ||
BC031456.1 | - | 10 | 1879 | 4037515 | |
BC064466.1 | - | 10 | 1972 | 6825549 |