Gene Transcripts

Gene Symbol
Dyrk2
Entrez Gene ID
69181
Gene Name
dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2
Gene Aliases
1810038L18Rik
Chromosome Location
Chr. 10: 118855602 - 118889246 on Build GRCm38
UniGene ID
Mm.45565
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014390.2 NP_001014412.1 3 1630
GenBank mRNA AK149371.1 - 3 1867
AK155496.1 - 3 1880
BC085145.1 - 3 1630 6808145

View Full Product Details