Gene Transcripts

Gene Symbol
Sstr5
Entrez Gene ID
20609
Gene Name
somatostatin receptor 5
Gene Aliases
Smstr5, sst5
Chromosome Location
Chr. 17: 25489875 - 25514304 on Build GRCm38
UniGene ID
Mm.353282
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191008.1 NP_001177937.1 2 1389
NM_011425.3 NP_035555.1 3 1541
GenBank mRNA AK133609.1 - 3 1507
AK133767.1 - 2 1317
BC150805.1 - 2 1373 9087716

View Full Product Details