Gene Transcripts

Gene Symbol
Rilp
Entrez Gene ID
280408
Gene Name
Rab interacting lysosomal protein
Gene Aliases
AI835553, Gm857
Chromosome Location
Chr. 11: 75510094 - 75513168 on Build GRCm38
UniGene ID
Mm.41416
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029938.2 NP_001025109.1 8 1380
GenBank mRNA AK139016.1 - 8 1372
AK160517.1 - 6 1173
BC038146.1 - 4 698
BC051502.1 - 5 839

View Full Product Details