Gene Transcripts

Gene Symbol
Tmprss11c
Entrez Gene ID
435845
Gene Name
transmembrane protease, serine 11c
Gene Aliases
Tmprss13
Chromosome Location
Chr. 5: 86231481 - 86289308 on Build GRCm38
UniGene ID
Mm.359654
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030297.2 NP_001025468.1 5 499
GenBank mRNA AM260217.1 - 5 499
BC147157.1 - 5 480 9056205
BC147158.1 - 5 480 9056165

View Full Product Details