Gene Transcripts

Gene Symbol
Macrod2
Entrez Gene ID
72899
Gene Name
MACRO domain containing 2
Gene Aliases
1110033L15Rik, 2610107G07Rik, 2900006F19Rik
Chromosome Location
Chr. 2: 140395356 - 142392966 on Build GRCm38
UniGene ID
Mm.229466
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028387.1 NP_082663.1 4 2052
GenBank mRNA AK029184.1 - 4 2032

View Full Product Details