Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001276378.1 | NP_001263307.1 | 12 | 2362 | |
NM_001276379.1 | NP_001263308.1 | 10 | 2133 | ||
NM_020347.3 | NP_065080.1 | 10 | 2081 | ||
NR_075080.1 | 9 | 1956 | |||
XM_006713207.2 | XP_006713270.2 | 9 | 1962 | ||
XM_011533838.2 | XP_011532140.1 | 10 | 2005 | ||
XM_017006644.1 | XP_016862133.1 | 13 | 2719 | ||
XM_017006645.1 | XP_016862134.1 | 12 | 2594 | ||
GenBank mRNA | AJ297351.1 | - | 10 | 2038 | |
AK027165.1 | - | 1 | 709 | ||
AK129805.1 | - | 1 | 125 | ||
BC042483.1 | - | 10 | 2041 | 4831113 | |
BX640604.1 | - | 10 | 2063 |