Gene Transcripts

Gene Symbol
Nrbf2
Entrez Gene ID
641340
Gene Name
nuclear receptor binding factor 2
Gene Aliases
NRBF-2
Chromosome Location
Chr. 10: 67266689 - 67285347 on Build GRCm38
UniGene ID
Mm.136223
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001036293.2 NP_001031370.1 4 1543
GenBank mRNA AK002810.1 - 4 1542
AK004200.1 - 4 1463
BC019448.1 - 4 1543 4212132

View Full Product Details