Gene Transcripts

Gene Symbol
TMSB15A
Entrez Gene ID
11013
Gene Name
thymosin beta 15a
Gene Aliases
TMSB15, TMSB15B, TMSL8, TMSNB, Tb15, TbNB
Chromosome Location
Chr. X: 102513682 - 102516771 on Build GRCh38
UniGene ID
Hs.56145
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021992.2 NP_068832.1 3 291
GenBank mRNA AF086495.1 - 3 234
BC000183.1 - 3 239 3350388
BG829126.1 - 3 241
BU599402.1 - 3 304
D82345.1 - 3 265

View Full Product Details