Gene Transcripts

Gene Symbol
OR8G2
Entrez Gene ID
26492
Gene Name
olfactory receptor family 8 subfamily G member 2
Gene Aliases
HSTPCR120, OR8G2P, OR8G4, ORL206, ORL486, TPCR120
Chromosome Location
Chr. 11: 124224639 - 124225663 on Build GRCh38
UniGene ID
Hs.381319
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291438.1 NP_001278367.1 1 673
GenBank mRNA BC136638.1 - 1 673
BC136639.1 - 1 673 9020629
X89669.1 - 1 206

View Full Product Details