Gene Transcripts

Gene Symbol
MAPKAP1
Entrez Gene ID
79109
Gene Name
mitogen-activated protein kinase associated protein 1
Gene Aliases
JC310, MIP1, SIN1, SIN1b, SIN1g
Chromosome Location
Chr. 9: 125437394 - 125707234 on Build GRCh38
UniGene ID
Hs.495138
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006617.1 NP_001006618.1 7 1271
NM_001006618.1 NP_001006619.1 7 1271
NM_001006619.1 NP_001006620.1 7 1271
NM_001006620.1 NP_001006621.1 6 943
NM_001006621.1 NP_001006622.1 5 853
NM_024117.3 NP_077022.1 7 1271
XM_011519005.1 XP_011517307.1 8 1412
XM_011519006.2 XP_011517308.1 8 1412
XM_011519007.1 XP_011517309.1 8 1412
XM_011519008.1 XP_011517310.1 8 1412
XM_011519009.1 XP_011517311.1 8 1412
XM_011519010.2 XP_011517312.1 4 589
XM_017015126.1 XP_016870615.1 7 1160
XM_017015127.1 XP_016870616.1 7 1160
GenBank mRNA AK290060.1 - 7 1084
AK295364.1 - 4 577
AK298841.1 - 3 345
AK309507.1 - 4 538
AK310361.1 - 4 577
AK316149.1 - 4 516
AL833042.1 - 6 904
AY524429.1 - 7 1268
AY524430.1 - 7 1268
AY524431.1 - 7 1268
AY524432.1 - 7 1268
AY633624.1 - 7 1238
AY633625.1 - 7 1245
AY633626.1 - 7 1245
BC002326.2 - 7 1240 2823015
BC003044.1 - 7 1240 2823015
M37191.1 - 6 716

View Full Product Details