Gene Transcripts

Gene Symbol
KLHDC9
Entrez Gene ID
126823
Gene Name
kelch domain containing 9
Gene Aliases
KARCA1
Chromosome Location
Chr. 1: 161098361 - 161100348 on Build GRCh38
UniGene ID
Hs.507290
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007255.2 NP_001007256.1 4 1246
NM_152366.4 NP_689579.3 4 1227
NR_033385.1 5 996
NR_033386.1 5 1015
GenBank mRNA AK093268.1 - 1 1855
AY601914.1 - 4 1245
BC022077.1 - 4 1226 4822843
BC032482.1 - 1 1884 5222141
BC066896.1 - 4 1221 4828667

View Full Product Details