Gene Transcripts
- Gene Symbol
- SMA4
- Entrez Gene ID
- 11039
- Gene Name
- glucuronidase beta pseudogene
- Gene Aliases
- SMA3, b55C20.2
- Chromosome Location
- Chr. 5: 70127462 - 70290177 on Build GRCh38
- UniGene ID
- Hs.582500
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029426.1 |
|
8 |
908 |
|
GenBank mRNA |
AK097091.1 |
- |
9 |
1288 |
|
Z70701.1 |
- |
2 |
102 |
|
- Gene Symbol
- GUSBP2
- Entrez Gene ID
- 387036
- Gene Name
- glucuronidase, beta pseudogene 2
- Gene Aliases
- GUSBL1, GUSBP4, SMA3-L, SMAC3L, SMAC3L2, b55C20.1, bA239L20.1, bA239L20.5, bGLU-Lp
- Chromosome Location
- Chr. 6: 26871487 - 26956554 on Build GRCh38
- UniGene ID
- Hs.675815
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC067351.1 |
- |
5 |
994 |
|
- Gene Symbol
- LOC653080
- Entrez Gene ID
- 653080
- Gene Name
- glucuronidase beta pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 5: 70785437 - 70795916 on Build GRCh38
- UniGene ID
- Hs.582500
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
Z70705.1 |
- |
1 |
386 |
|
View Full Product Details