Gene Transcripts

Gene Symbol
SLC22A25
Entrez Gene ID
387601
Gene Name
solute carrier family 22 member 25
Gene Aliases
HIMTP, UST6
Chromosome Location
Chr. 11: 63162264 - 63243523 on Build GRCh38
UniGene ID
Hs.332119
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199352.3 NP_955384.3 1 299
XM_017017684.1 XP_016873173.1 4 1378
XM_017017685.1 XP_016873174.1 5 1506
XM_017017686.1 XP_016873175.1 4 1814
XM_017017687.1 XP_016873176.1 4 1919
XM_017017695.1 XP_016873184.1 4 1919
GenBank mRNA AK127610.1 - 1 174
AY437532.1 - 1 299
BC101315.1 - 1 294
BC101316.1 - 1 299
BC101317.1 - 1 299

View Full Product Details