Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020664.3 | NP_065715.1 | 9 | 1564 | |
GenBank mRNA | AJ293009.1 | - | 9 | 1479 | |
AK094340.1 | - | 4 | 2048 | ||
AK225961.1 | - | 9 | 1733 | ||
AL442076.1 | - | 2 | 691 | ||
BC010740.1 | - | 9 | 1550 | 3877040 | |
BC011968.1 | - | 9 | 1469 | 3866503 |