Gene Transcripts

Gene Symbol
Vom1r29
Entrez Gene ID
494292
Gene Name
vomeronasal 1 receptor 29
Gene Aliases
V1re10
Chromosome Location
Chr. 1: 68221395 - 68222312 on Build Rnor_6.0
UniGene ID
Rn.129437
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008946.1 NP_001008946.1 1 210
GenBank mRNA AY510346.1 - 1 210

View Full Product Details