Gene Transcripts

Gene Symbol
Vom1r88
Entrez Gene ID
494233
Gene Name
vomeronasal 1 receptor 88
Gene Aliases
V1rc28
Chromosome Location
Chr. 4: 88423412 - 88424305 on Build Rnor_6.0
UniGene ID
Rn.129652
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009513.1 NP_001009513.1 1 535
GenBank mRNA AY510324.1 - 1 535

View Full Product Details